Would you like to log in again?
Maxwell® 16 LEV RNA FFPE Purification Kit, Promega
Supplier: Promega
Maxwell®
AS1260
PAAS1260EA 667.95 USD InStock
PAAS1260
Maxwell® 16 LEV RNA FFPE Purification Kit, Promega
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
The Maxwell 16 LEV RNA FFPE Purification Kit is designed to extract RNA rapidly from FFPE samples using the Maxwell 16 Instrument.
  • Automated Platform for Extracting RNA from FFPE Mammalian Tissues
  • Walkaway Automated Extraction from up to 16 Samples in a Single Run
  • Pure and Amplifiable RNA from FFPE Mammalian Tissue Samples
  • Simple Method with No Organic Solvents for Deparaffinization
  • Decreased Processing Time
The FFPE kit is specifically designed for optimal purification of RNA from 5 μm-thin sections of formalin-fixed, paraffin-embedded (FFPE) tissue samples
This kit does not use hazardous xylene, thereby providing a much safer method than many competitive kits. Quality testing demonstrates virtually no PCR inhibitors in purified samples.
The Maxwell 16 LEV RNA FFPE Purification Kit is optimized to yield a final RNA concentration that maximizes RNA yield, eliminating the need to concentrate the extract prior to amplification. Lysates are placed directly into the cartridges, and RNA ready for amplification is obtained in approximately 60 minutes (after proteinase K digest).
Accessories information: Maxwell 16 LEV Cartridge Rack Available Separately, One Each.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty