Would you like to log in again?
TBE Buffer, 10X, Molecular Biology Grade, Promega
Supplier: Promega
V4251
PAV4251EA 83.47 USD InStock
PAV4251
TBE Buffer, 10X, Molecular Biology Grade, Promega
Buffers Electrophoresis Buffers Running Buffers
TBE Buffer, 10X (pH 8.3), is used for polyacrylamide and agarose gel electrophoresis.
  • Quality tested; demonstrates ≤1% release of DNase and RNase activity
This product has been optimized for use in DNA applications.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty