Would you like to log in again?
Difco™ Letheen Agar, BD Diagnostics
Supplier: BD Diagnostics
268010 268010
90003-798EA 364.28 USD InStock
90003-798 CA90003-798
Difco™ Letheen Agar, BD Diagnostics
Media for Microbiology Prepared Media for Microbiology
For evaluating the bactericidal activity of quaternary ammonium compounds.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty