Would you like to log in again?
PNGase F
Supplier: Promega
V4831
PAV4831EA 214.58 USD InStock
PAV4831
PNGase F
Enzymes
PNGase F is a recombinant glycosidase cloned from Elizabethkingia miricola and overexpressed in E. coli. PNGase F has a molecular weight of 36kDa.
  • Recombinant Glycosidase Cloned from Elizabethkingia miricola
  • Use to determine protein glycosylation status and location
  • Cleaves N-linked oligosaccharides between innermost GlcNAc and asparagines from N-linked glycoproteins
  • Will not remove oligosaccharides found on plant glycoproteins
  • Supplied at a concentration of 10u/microl
PNGase F is a recombinant glycosidase cloned from Elizabethkingia miricola and overexpressed in E. coli. PNGase F has a molecular weight of 36kDa. PNGase F catalyzes the cleavage of N-linked oligosaccharides between the innermost GlcNAc and asparagine residues of high mannose, hybrid and complex oligosaccharides from N-linked glycoproteins. PNGase F will not remove oligosaccharides containing Alpha-(1,3)-linked core fucose commonly found on plant glycoproteins. Unit Definition: One unit of PNGase F will catalyze the deglycosylation of 1 nanomole of denatured Ribonuclease B (RNase B) in one minute at 37°C. One Promega unit is equal to 1 IUB milliunit. Molecular Weight: PNGase F has a molecular weight of approximately 36kDa. Physical Form: PNGase F is supplied as a liquid in 20mM Tris-HCl (pH 7.5 at 25°C), 50mM NaCl and 5mM EDTA at a concentration of 10,000u/ml.
One Promega unit is equal to one IUB milliunit. PNGase F has a molecular weight of approximately 36 kDa. PNGase F is supplied as a liquid in 20 mM Tris-HCl (pH 7.5 at 25°C), 50 mM NaCl, and 5mM EDTA at a concentration of 10,000 u/mL.
PNGase F does not remove oligosaccharides containing Alpha-(1,3)-linked core fucose commonly found on plant glycoproteins. One unit of PNGase F will catalyze the deglycosylation of one nanomole of denatured Ribonuclease B (RNase B) in one minute at 37°C.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty