Would you like to log in again?
IdeS Protease
Supplier: Promega
V7515 V7511
PAV7511EA 1201.69 USD InStock
PAV7511 PAV7515
IdeS Protease
Enzymes
IdeS Protease is an immunoglobulin-degrading enzyme from Streptococcus pyogenes (IdeS). IdeZ Protease is an immunoglobulin-degrading enzyme from Streptococcus equi subspecies zooepidemicus.
  • Antibody Fragmentation and Characterization, Simplified
  • Highly specific IgG cleavage
  • Rapid generation of highly pure fragments
  • Frozen protease is supplied at a concentration of 50u/µl
IdeS Protease: IdeS Protease is an immunoglobulin-degrading enzyme from Streptococcus pyogenes (IdeS). It is an engineered recombinant protease overexpressed in E. coli that cleaves Immunoglobulin G (IgG) with high specificity at a single site below the hinge region, yielding F(ab’)2 and Fc fragments. The protocol for a standard reaction is to add the IdeS Protease to the IgG sample, add 1 unit of IdeS Protease per 1ug of IgG to be digested and incubate the sample at 37°C for 30-60 minutes in a neutral pH buffer. IdeZ Protease: IdeZ Protease is an immunoglobulin-degrading enzyme from Streptococcus equi subspecies zooepidemicus. It is an engineered recombinant protease overexpressed in E. coli. Like IdeS Protease, IdeZ Protease specifically cleaves IgG molecules below the hinge region to yield F(ab′)2 and Fc fragments. However, IdeZ Protease has significantly improved activity against mouse IgG2a and IgG3 subclasses compared to IdeS Protease.
IdeZ Protease is an engineered recombinant protease overexpressed in E. coli. Like IdeS Protease, IdeZ Protease specifically cleaves IgG molecules below the hinge region to yield F(ab′)2 and Fc fragments. However, IdeZ Protease has significantly improved activity against mouse IgG2a and IgG3 subclasses compared to IdeS Protease.
coli</italic> that cleaves Immunoglobulin G (IgG) with high specificity at a single site below the hinge region, yielding F(ab’)2 and Fc fragments. The protocol for a standard reaction is to add the IdeS Protease to the IgG sample, add one unit of IdeS Protease per 1 µg of IgG to be digested, and incubate the sample at 37°C for 30–60 minutes in a neutral pH buffer.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty