Would you like to log in again?
Transfer Racks for ThermoMixers, Eppendorf
Supplier: Eppendorf
3880000151 3880000151 3880000305 3880000305
75856-502EA 75.88 USD InStock
CA75856-502 75856-502 CA75856-500 75856-500
Transfer Racks for ThermoMixers, Eppendorf
Racks Tube Racks
These transfer racks are designed as a work rack for sample preparation and used to maintain accurate and uniform temperatures.
  • Individually sensor-controlled
  • Holds up to 24 microtubes for easy transfer of multiple vessels in parallel
  • Convenient transfer of up to 24 hot vessels without burning the fingers
  • Temperature range from ‒96 to 121 °C for broad flexibility
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty