Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hyaluronic acid sodium salt (from Streptococcus equi), high viscosity, Calbiochem®

Supplier: MilliporeSigma
Synonyms: HA sodium saltSodium hyaluronate

385908-500MG
80056-712EA 347.36 USD InStock
80056-712
Hyaluronic acid sodium salt (from Streptococcus equi), high viscosity, Calbiochem®
Hyaluronic acid sodium salt
Natural high-viscosity mucopolysaccharide with alternating β1,3-glucuronidic and β1,4-glucosaminidic bonds. Principal glycosaminoglycan in connective tissue fluids.

The molecular weight of Hyaluronic Acid, Sodium Salt, is in the range of 5.0 x 105 - 1.2 x 106, as calculated from its limiting viscosity.


Formula: (C₁₄H₂₁NaNO₁₁)n
Storage Temperature: Refrigerator
MDL Number: MFCD00875848
CAS Number: 9067-32-7
Order Now
Specification Test Results
Form Lyophilized
Hygroscopic Hygroscopic
Contaminants protein: ≤0.1% endotoxins: ≤ 0.070 EU/mg NaHy
Toxicity Standard Handling
Solubility H₂O (10 mg/ml). Heating slowly to 50°C may be required for complete solubilization.
Storage +2 °C - +8 °C (Hygroscopic)
Do not freeze Ok to freeze

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty