Would you like to log in again?
SP Wilmad-LabGlass Vacuum Hydrolysis Tubes, SP Industries
Supplier: SP Industries
LG-10803-100
14207-868EA 237.49 USD InStock
14207-868
SP Wilmad-LabGlass Vacuum Hydrolysis Tubes, SP Industries
Tubes Test Tubes
These borosilicate glass tubes include a valve plug with plastic knob.
  • Designed for sealed tube reactions
High temperatures can be generated if used with an aluminum block to transfer heat directly to the glass tube. Applications include sample concentration, protein hydrolysis, and freeze drying.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty