Would you like to log in again?
Vernier Projectile Stop
Supplier: Vernier
PS-VPL
470223-686EA 39.99 USD InStock
470223-686
Vernier Projectile Stop
Educational Materials Physics Educational Materials Mechanics Learning Activities
Control Your Projectiles!
  • The Projectile Stop is heavy foam-coated steel that stays where you put it
The Projectile Stop has one job–to keep the projectiles from the Vernier Projectile Launcher from rolling out of sight. Place the Projectile Stop in the launch line, beyond the landing site, and the projectile will be caught.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty