Would you like to log in again?
3B Scientific® Rat Brain
Supplier: AMERICAN 3B SCIENTIFIC SE
3B Scientific®
1000230
470224-890EA 398.6 USD InStock
470224-890
3B Scientific® Rat Brain
Models Zoology Models
This model shows a rat brain in approx. 6-fold enlargement. Sectioned medially, it can be disassembled into two halves.
  • Base includes a diagram of the human brain for comparison
  • 3 year manufacturers warranty
The right half of the model shows the structures of the cerebrum, cerebellum and brain stem, each of which is color-coded for didactic purposes (cerebrum = pink, cerebellum = blue, brain stem = yellow), both externally and in the median section.
The left half of the model is largely transparent, thus revealing a view of the colored left lateral ventricle and hippocampus, which can also be seen in the median section. For purposes of comparison, a natural cast of a rat brain and a didactic, small-scale illustration of a human brain in median section are shown on the base, with the same color coding used for the various regions.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty