Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Perchloric acid concentrate for dilution to 1L, 0.1 M HClO4 (0.1N) volumetric solution, for titration in non-aqueous media, Fluka™

Danger

32046-1EA
BJ32046-1EAEA 116.46 USD InStock
BJ32046-1EA
Perchloric acid concentrate for dilution to 1L, 0.1 M HClO4 (0.1N) volumetric solution, for titration in non-aqueous media, Fluka™
Perchloric acid
concentrate for 1L standard solution


Formula: HClO₄
MW: 100.46 g/mol
Boiling Pt: 203 °C
Melting Pt: -18 °C
Density: 1.66 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011325
CAS Number: 7601-90-3
UN: 1873
ADR: 5.1,I
https://us-legacy.vwr.com/stibo/web/std.lang.all/06/22/16640622.jpg
Order Now
Specification Test Results
Factor 0.998 - 1.002

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty