Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ammonium thiocyanate 0.1 M NH4SCN (0.1N) volumetric solution, Fluka™

Supplier: Honeywell Research Chemicals
Synonyms: Ammonium rhodanide

35036-1L
BJ35036-1LEA 42.81 USD InStock
BJ35036-1L
Ammonium thiocyanate 0.1 M NH4SCN (0.1N) volumetric solution, Fluka™
Ammonium thiocyanate


Formula: NH₄SCN
MW: 76.12 g/mol
Boiling Pt: 100 °C (1.013 hPa)
Storage Temperature: Ambient
MDL Number: MFCD00011428
CAS Number: 1762-95-4
EINECS: 231-791-2
Order Now
Specification Test Results
Factor 0.998 - 1.002

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty