Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

n-Octane ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

050800100MG
100274-320EA 111.15 USD InStock
100274-320
n-Octane ≥99%
n-Octane
Storage conditions: Store in a cool, dry place


Formula: H₃C(CH₂)₆CH₃
MW: 114.23 g/mol
MDL Number: MFCD00009556
CAS Number: 111-65-9
UN: 1262
ADR: 3,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/61/84/21286184.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty