Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Pentanol ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS
Synonyms: 1-Pentyl alcoholn-Amyl alcoholAmyl alcohol1-Hydroxy pentaneButyl carbinol

400500100MG
100298-760EA 196.15 USD InStock
100298-760
1-Pentanol ≥99%
1-Pentanol
Storage conditions: Store in a cool, dry place


Formula: H₃C(CH₂)₃CH₂OH
MW: 88.15 g/mol
MDL Number: MFCD00002977
CAS Number: 71-41-0
UN: 1105
ADR: 3,III
https://us-legacy.vwr.com/stibo/web/std.lang.all/71/16/21287116.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty