Would you like to log in again?
Difco™ Lennox Agar, BD Diagnostics
Supplier: BD Diagnostics
240110 240110
90003-112EA 241.57 USD InStock
90003-112 CA90003-112
Difco™ Lennox Agar, BD Diagnostics
Media for Microbiology Prepared Media for Microbiology
For propagation and maintenance of E.coli for molecular biology.
  • Contains 5g/L sodium chloride
  • Dehydrated culture media
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty