Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Whatman™ Anotop Plus Syringe Filters, Prefilter, Sterile, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
Anotop™ Whatman™ Anotop™
6809-3122 6809-3112 6809-3112 6809-3122 6809-3102 6809-3102 6809-4112 6809-4122 6809-4102 6809-4122 6809-4102 6809-4112
28138-033PK 1051.12 USD InStock
28138-033 CA28138-051 28138-047 28138-037 CA28138-033 CA28138-049 CA28138-047 28138-049 CA28138-037 CA10147-958 28138-051 10147-958
Whatman™ Anotop Plus Syringe Filters, Prefilter, Sterile, Whatman products (Cytiva)
Filters Syringe Filters
Cytiva's Whatman Anotop Plus sterile syringe filter with glass microfiber prefilter is compatible with most organic and aqueous solutions for use in a wide variety of applications.
  • Contains an inorganic Anopore membrane and is available in three pore sizes
  • Polypropylene syringe filters comes in two diameters depending on sample volume
  • Integrated glass microfiber prefilter aids in filtration of difficult samples
  • Recyclable after use
Anotop Plus filters are manufactured with an Anopore membrane suitable for many applications, including capillary electrophoresis, UV/Vis sample preparation, protein separation, and HPLC sample preparation.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty