Would you like to log in again?
TrueBlue™ Peroxidase Substrate, KPL
Supplier: LGC Clinical Diagnostics
5510-0030 5510-0052 5510-0049
95059-168EA 364.99 USD InStock
95059-168 95059-470 95059-468
TrueBlue™ Peroxidase Substrate, KPL
Assays Immunological Assays
TrueBlue is a highly sensitive chromogenic substrate for visualization of horseradish peroxidase-labeled reporter reagents
  • Free of known carcinogens
  • Recommended for detection of the least abundant marker
  • High sensitivity
TrueBlue is up to 100 times more sensitive than DAB and it is free of known carcinogens. Strong blue reaction product provides excellent contrast with DAB and other substrates. HistoMark TrueBlue provides the most sensitive means available to detect peroxidase-labeled conjugates. Because of its high sensitivity, TrueBlue is recommended for detection of the least abundant marker. TrueBlue is alcohol and xylene insoluble.
It provides a brilliant blue specific stain, which contrasts sharply with red counterstains. TrueBlue Substrate is a buffered solution containing 3, 3’, 5, 5’-tetramethylbenzidine (TMB) and Hydrogen Peroxide (H2O2).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty