Would you like to log in again?
FluorSave™ Reagent, MilliporeSigma
Supplier: MilliporeSigma
Calbiochem®
345789-20ML
EM345789-20MLEA 159.33 USD InStock
EM345789-20ML
FluorSave™ Reagent, MilliporeSigma
Cell Culture Supplements and Additives
FluorSave™ Reagent is an aqueous mounting medium for use in conjunction with fluorescent tracers such as fluorescein, rhodamine, and phycobiliprotein probes.
  • Preserves fluorescence-stained tissue sections
  • Ideal for use with phycobiliprotein tracers
  • Specially formulated without glycerol
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty