Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Water, Molecular Biology Grade, sterile RNase Free, DNase Free, Protease Free, DEPC Free

Supplier: Ricca Chemical

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Synonyms: Dihydrogen oxide

R9145000-1G
75799-280CS 661.61 USD InStock
75799-280
Water, Molecular Biology Grade, sterile RNase Free, DNase Free, Protease Free, DEPC Free
Water
Suitable for molecular and cell biology specifications.

  • RNase, DNase, Protease, Endotoxin-Free
  • DEPC Free
  • Sterile


Formula: H₂O
MW: 18.02 g/mol
Boiling Pt: 100 °C
Melting Pt: 0 °C
Density: 0.997 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
EINECS: 231-791-2
Merck Index: 12,10175
Order Now
Specification Test Results
Appearance Clear, colorless liquid
Bioburden (no growth) To Pass Test
DNase Activity (none detected) To Pass Test
Endotoxin (none detected) To Pass Test
Protease Activity (none detected) To Pass Test
RNase Activity (none detected) To Pass Test

 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty