Would you like to log in again?
EnzyChrom™ Alanine Assay Kit, BioAssay Systems
Supplier: BioAssay Systems
EALA-100
75878-036EA 582.45 USD InStock
75878-036
EnzyChrom™ Alanine Assay Kit, BioAssay Systems
Assays Immunological Assays
For quantitative determination of L-Alanine and evaluation of drug effects on alanine metabolism.
  • Linear detection range 1 to 200 μM alanine for colorimetric assays and 0.4 to 20 μM for fluorimetric assays
Alanine, a nonessential amino acid, is utilized in the glucose-alanine cycle between tissues and the liver
BioAssay Systems’ Alanine Assay Kit provides a simple, direct and automation-ready procedure for measuring alanine concentration. Alanine is converted into pyruvate which can then be directly measured. The color intensity of the reaction product at 570nm or fluorescence intensity at λex/em = 530/585nm is directly proportional to the alanine concentration in the sample.
In tissues that metabolize amino acids, amino groups are collected as glutamate by transamination. The amine group is then transferred by alanine transaminase (ALT) from glutamate to pyruvate to form alanine and a-ketoglutarate. The alanine generated is transported to the liver where a reverse ALT reaction occurs and pyruvate is regenerated. Pyruvate is converted through gluconeogenesis to glucose which can then be recirculated to the tissues. Alanine concentration may have some correlation with high blood pressure, energy intake, cholesterol levels and body mass index.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty