Would you like to log in again?
ErgoDeck® Comfort Workstation Floor Mat Kitl, Wearwell®
Supplier: Wearwell
ErgoDeck®
556.78X42X48CHYL 715411005307 715411003990
75876-248EA 969.31 USD InStock
75876-248 75876-322 75876-320
ErgoDeck® Comfort Workstation Floor Mat Kitl, Wearwell®
Floor Mats Traction and Anti-Fatigue Floor Mats
The ErgoDeck® Comfort Kit contains all of the components for a workstation mat and assembles in 5 minutes.
  • Standing relief in wet or dry applications
  • Kit contains all of the components to create a workstation mat
  • Assembles in 5 minutes
  • Positive interlocking system prevents separation
  • 100% Silicone-free PVC tiles provide maximum durability
ErgoDeck® Comfort provides “rubber-like” resilience underfoot with its spring-back design. It is engineered to cushion the impact of long-term standing, and is ideally suited to work in areas that remain relatively dry, or have slight overspray. The raised surface design delivers substantial traction and 6" wide borders on three sides prevents trips.
Certifications: ASTM F1677. Dry: COF = 0.65. Wet: COF = 0.42.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty