Would you like to log in again?
Arginase Inhibitor Screening Kit, BioAssay Systems
Supplier: BioAssay Systems
IARG-100
76121-352EA 365.02 USD InStock
76121-352
Arginase Inhibitor Screening Kit, BioAssay Systems
Assays Immunological Assays
HTS for inhibitor screening and evaluation of arginase inhibitors.
  • Non-radioactive assay.
  • High-throughput. Homogenous “mix-incubate-measure” type assay. Can be readily automated on HTS liquid handling system
  • Rapid and reliable. Can be completed in less than 2 hours and no 37°C heater is needed.
  • Method: OD430nm
  • Samples: Arginase
ARGINASE (L-arginine ureohydrolase EC 3.5.3.1) is present in mammals and plants. In humans, arginase is expressed predominantly in the liver, and to lesser degrees in breast, kidney, testes, salivary glands, epidermis and erythrocytes. Arginase catalyzes the conversion of arginine to ornithine and urea, important for protection against NH3 toxicity and for cell growth and repair. Excessive arginase activity has been linked to cardiovascular diseases, also contribute to vascular structural problems and neural toxicity. Studies show that arginase inhibitors have been proved to be beneficial in cardiovascular and nervous system diseases. Simple, direct and automation-ready procedures for measuring arginase inhibition are highly desirable in Research and Drug Discovery. BioAssay Systems' arginase inhibitor screening kit provides a sensitive and convenient method to screen for arginase inhibitors. The method utilizes a chromogen that forms a colored complex specifically with urea produced in the arginase reaction. The intensity of the color is directly proportional to the arginase activity in the sample. Percent inhibition of a test compound can be determined by comparing the color intensity of the reaction preincubated with the test compound with the color intensity of an untreated control reaction
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty