Would you like to log in again?
Silver Capsules, CE Elantech
Supplier: CE Elantech
240-054-00 240-054-05
101189-550PK 48.46 USD InStock
101189-550 101189-548
Silver Capsules, CE Elantech
Sample Containers General Purpose Containers
These Silver Soft containers are obtained by pressing light metal foil and are ideal for solid samples
These are recommended for Total Organic Carbon (TOC) determination.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty