Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium orthovanadate

Supplier: BIOASSAY SYSTEMS

PHIVA-1ML
75878-184EA 171.2 USD InStock
75878-184
Sodium orthovanadate
Sodium orthovanadate
A general inhibitor for protein phosphotyrosyl phosphatases.

  • Ready-to-use phosphatase inhibitor
  • Fully activated for maximum phosphatase inhibition

Sodium orthovanadate (CAS Number 13721-39-6, Na3VO4) is a general inhibitor for protein phosphotyrosyl phosphatases (PTPs)

BioAssay Systems’ Vanadate reagent is activated for maximum inhibition. Suggested Working Concentration: 1-10 mM

Inhibition of vanadate is competitive and can be reversed by addition of EDTA or by dilution. Vanadate is commonly used to preserve the protein tyrosyl phosphorylation state in cells and lysates.


Formula: Na₃O₄V
MW: 183.91 g/mol
MDL Number: MFCD00003511
CAS Number: 13721-39-6
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty