Would you like to log in again?
Hi-Voltage ARC™ Safety Glasses, Protective Industrial Products
Supplier: Protective Industrial Products
250-24-0000
89179-578CS 727.04 USD InStock
89179-578
Hi-Voltage ARC™ Safety Glasses, Protective Industrial Products
Safety Glasses
Eye protection with a semi-rimless frame and built-in sideshields.
  • U-Fit® spatula temples
  • 4 base curve lens
  • Lens design maximizes view, providing user comfort and safety
  • Adjustable length temples provide a secure and comfortable fit
  • Molded nose bridge design to fit the vast majority of users
Clear, crisp, and distortion-free visibility while meeting the requirement standards that help provide protection from projectiles and impact in general use applications. Treated front and back with a clear, scratch-resistant coating for a much harder surface.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty