Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IgA Goat Polyclonal Antibody

Supplier: MP Biomedicals
0855167
75877-358EA 463 USD InStock
75877-358
Anti-IgA Goat Polyclonal Antibody
Antibodies
Product is the lyophilized powder of goat antiserum to human secretory IgA and buffer salts.

IgA antibody is a secretory antibody found abundantly in mucous linings of gastrointestinal, respiratory and genitourinary tracts, tears and saliva. Secretory IgA contributes to the humoral defense mechanism against the pathogens on mucosal surfaces. Anti-Human IgA (α-chain specific) antibody specifically recognizes human IgA. Immunoglobulin A (IgA) is the primary antibody involved in mucosal immunity. It is identified by the presence of α-type heavy chains. IgA, which is composed of two subclasses IgA1 and IgA2, can exist in a dimeric secretory form found in mucous secretions.

The total protein is measured using the Biuret procedure, with bovine albumin as a standard. Antibody titer is standardized by equivalence-point precipitation. Each antiserum is tested for specificity at a minimum of 80 mg/mL using immunoelectrophoresis.

The antiserum is prepared by hyperimmunizing goats with highly purified human secretory IgA; antiserum is delipidated and made specific by solid-phase absorption. The resulting antiserum is dialyzed into 0.02M sodium phosphate, 0.14M sodium chloride, pH 7.3, with 0.05% sodium azide, adjusted to standard titer, filtered through a 0.22 µm filter, vialed and lyophilized.

This product is suitable for use as a precipitating antibody. It gives sharp precipitin lines in immunoelectrophoresis and immunodiffusion techniques. It is suitable for Ouchterlony double diffusion. The antibody was used for IgA quantification by ELISA in various studies. Antibody dilution of 1:4000 was used for passive hemagglutination inhibition assay.

Type: Secondary
Antigen: IgA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty