Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IgG Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: MP Biomedicals
0855689
75877-488EA 465.76 USD InStock
75877-488
Anti-IgG Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Peroxidase-conjugated goat affinity purified antibody to rabbit IgG (whole molecule) is horseradish peroxidase (HRP)-conjugated goat affinity purified antibody to rabbit IgG (whole molecule) and buffer salts. Antiserum is produced in goat using purified rabbit IgG as the immunogen. Antibody is isolated from goat anti-rabbit IgG antiserum by immunospecific purification which removes essentially all goat serum proteins, including immunoglobulins, which do not specifically bind to rabbit IgG. The antibody preparation is solid phase adsorbed with human IgG to ensure minimal cross reactivity in tissue or cell preparations.

Specificity of the Anti-Rabbit IgG-Peroxidase is determined by immunoelectrophoresis (IEP) using normal rabbit serum and rabbit IgG. The conjugate shows no reaction with human IgG by IEP. Identity and purity of the antibody is established by immunoelectrophoresis prior to conjugation. Electrophoresis of the antibody preparation followed by diffusion versus anti-goat IgG and anti-goat whole serum results in single arcs of precipitation.

Peroxidase-conjugated goat affinity purified antibody to rabbit IgG (whole molecule) is suitable for use as a reagent in enzyme immunoassays (EIA), cell and tissue staining (for light microscopy), cell and tissue labeling (for electron microscopy), and blot immunostaining. Use of affinity purified antibodies is recommended to avoid non-specific binding from the inherent antibodies of host animals. Note: F(ab')2 fragments are recommended for staining of cells or tissues which contain Fc receptors.

Antibody and highly purified HRP (Rz>3.0) are conjugated under defined conditions to obtain optimally labeled product. Conjugated protein is purified by salt fractionation. The product is dialyzed into 0.02M sodium phosphate, 0.14M sodium chloride, pH 7.3, with 1% BSA, adjusted to standard titer, filtered through a 0.22 µm filter, vialed, and lyophilized. No preservative is added.

The total protein is measured using the Biuret procedure with bovine albumin as standard. To ensure that the product titer falls within the required range, antibody titer is standardized by microtiter plate ELISA with rabbit IgG. The product is tested for purity and specificity at final concentration by immunoelectrophoresis. The antibody is goat IgG; no trace of albumin is detected.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype:
Reactivity: Rabbit

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty