Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cold Traps, Yamato

Supplier: Yamato
CA-301-115V CA-801-220V CA-801-115V
76550-224EA 8668.87 USD InStock
76550-224 76550-226 75846-430
Cold Traps, Yamato
Vacuum Traps
This space-saving unit traps water vapor and toxic substances discharged from the vacuum oven and rotary evaporator to protect the vacuum pump.

  • Direct trap method
  • Maximum low temperature: −45 °C/−85 °C
  • Coolant: R404A/R600a
  • Other applications: Low-temperature tank, pre-cooling tank

The smaller unit can dehumidify up to 0.9 kg of water-type liquid, and the larger unit, 1.0 kg.

A seven-segment LED screen displays the temperature. Safety features include an electric leakage breaker with overcurrent protection and refrigeration overload relay.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty