Would you like to log in again?
SP Wilmad-LabGlass Barrett Moisture Traps with 2 mm PTFE Plug, SP Industries
Supplier: SP Industries
LG-9076T-102
14204-752EA 787.55 USD InStock
14204-752
SP Wilmad-LabGlass Barrett Moisture Traps with 2 mm PTFE Plug, SP Industries
Distillation Components Distillation Receivers
Calibrated “to contain” at 20°C, these distilling receivers are used for determination of water in various materials by distillation with a solvent that is not miscible with water.
  • Borosilicate glass
With full length Standard Taper inner member for connection to distilling flask and a Standard Taper outer joint of the same size to connect the condenser, these moisture traps are supplied with a 2 mm PTFE stopcock.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty