Would you like to log in again?
Vacufuge Plus Concentrator, Eppendorf
Supplier: Eppendorf
022822993
75875-188EA 14404.45 USD InStock
75875-188
Vacufuge Plus Concentrator, Eppendorf
Concentrators
The Eppendorf Vacufuge plus assures quick, efficient and gentle vacuum concentration of DNA/RNA, nucleotides, proteins and other liquid or wet samples. It fits comfortably right on a lab bench.
  • Three system configurations available
  • Quiet operation <50 dB(A)
  • PTFE diaphragm pump eliminates the need for changing pump oil
  • Small footprint
  • 15 rotor options accommodate tubes from 0.2 mL to 50 mL, microplates and PCR plates
  • Can be connected with most commercially available cold traps for use with vapors
  • Coated lid with superior chemical resistance
  • Brake function
  • Start/Stop of concentration process
The Vacufuge plus was designed with the customer in mind to deliver an ergonomic, intuitive and durable solution that fits comfortably right on the lab bench
The Vacufuge plus is available with three configurations to meet the customers' lab-specific needs. The Basic Vacufuge is for connecting to an existing vacuum pump. The Complete System includes an integrated PTFE pump and condensation trap. Pump is solvent-resistant and maintenance and oil-free. The third configuration, the Complete System with vacuum port, adds extra capability. It allows for using the integrated vacuum pump separately, without having to disconnect it from the system.
System features include timer selection between 1 min and 9:59 h, a mode/vent function which allows you to choose between three solvent modes, and temperature preselection of room temperature, 30°C, 45°C, and 60°C. Also features a motorless induction drive and chemical-resistant, stainless steel chamber.
An advanced heating technology provides best treatment for your sample which assures quick, efficient and gentle vacuum concentration of DNA/RNA, nucleotides, proteins and other liquid or wet samples. The new coated lid provides superior chemical resistance against aggressive acids and organic solvents (e.g. TFA, DMSO).
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty