Would you like to log in again?
Magnetic Transfection Set, OZ Biosciences
Supplier: OZ Biosciences
IV-MAG2 IV-MAG3 IV-MAG1
103258-110EA 138.92 USD InStock
103258-110 103258-356 103258-108
Magnetic Transfection Set, OZ Biosciences
Magnets
Magnetic set for In vivo Magnetofection technology.
  • Can be easily cleaned and decontaminated with 70% ethanol, or by dry or wet sterilization
  • Can be used at room temperature, 37°C, +4°C, etc.
  • Solid, completely reusable, it is a one-time buy
Magnetic properties, distance between magnets and cells and incubation time have been optimized to efficiently concentrate nucleic acids or virus at the targeted site.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty