Would you like to log in again?
In vivo DogtorMag™ Transfection Reagent, OZ Biosciences
Supplier: OZ Biosciences
DogtorMag™
IV-TK30220 IV-KC30220 IV-DM30500 IV-DM31000
103258-282EA 908.66 USD InStock
103258-282 103258-272 103258-158 103258-280
In vivo DogtorMag™ Transfection Reagent, OZ Biosciences
Transfection Reagents
In vivo DogtorMag Transfection Reagent has been designed for in vivo targeted transfection of various types of nucleic acids such as DNA, RNA, oligonucleotides using magnetic nanoparticles coated with synthetic lipids.
  • Increased transfection efficiency
  • Targeted process (magnetically-driven)
  • Reduction of the systemic dissemination of vectors during injection
  • Reduction of the vector doses
  • Work under non permissive conditions (hypothermia, physiological flow conditions)
  • Penetration of the vector into tissues
  • Minimized toxicity
  • Universal - suitable for all nucleic acids
Accessories information: This reagent needs to be used with a magnetic plate.
Delivery information: Shipped at room temperature
Packaging: Store in vivo DogtorMag at -20 °C and in vivo CombiMag at +4 °C
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty