Would you like to log in again?
Masterclear® Self-Adhesive PCR Film, Eppendorf
Supplier: Eppendorf
0030132947
75875-068CS 367.94 USD InStock
75875-068
Masterclear® Self-Adhesive PCR Film, Eppendorf
Seals Microplate Seals
This semi-transparent film allows you to seal your real-time PCR plate leak tight.
  • Optimized for maximum light transmission
  • Optimized for tight sealing
  • Optimized packaging for your convenience
Use for PCR, real-time PCR, and other assays in plates.
A high transparency of over 90% between 450 nm and 750 nm guarantees maximum fluorescence excitation and read-out.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty