Would you like to log in again?
Electric Whirl
Supplier: EISCO SCIENTIFIC LLC
PH0904
470226-556EA 39 USD InStock
470226-556
Electric Whirl
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Demonstrate rotary motion from a charge.
  • For demonstrating the effect of electric discharge from points
The apparatus has a 95 mm diameter, Three-pointed star wheel with all the points curved in one direction. The wheel rotates freely on a pivot point mounted on a short stand and the stand is provided with a 4 mm connection socket.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty