Would you like to log in again?
Ab-DeliverIN™ Transfection Reagent, OZ Biosciences
Supplier: OZ Biosciences
Ab-DeliverIN™
AI21000 AI20250 AI20500 AI20100
103258-296EA 428.89 USD InStock
103258-296 103258-294 103258-300 103258-298
Ab-DeliverIN™ Transfection Reagent, OZ Biosciences
Transfection Reagents
Ab-DeliverIN Transfection Reagent is the first reagent dedicated to the intracellular delivery of fully functional antibodies.
  • Intracellular delivery of functionally active antibody
  • Highly efficient in many primary cells and cell lines
  • Serum compatible
  • Biodegradable and non toxic
  • Straightforward protocol and ready-to-use reagent
Delivery information: Shipped at room temperature
Packaging: Store Ab-DeliverIN at +4 °C and FITC labeled IgG at -20 °C
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty