Would you like to log in again?
HYPE-5™ Transfection Reagent, OZ Biosciences
Supplier: OZ Biosciences
HYPE-5™
HYR10003 HY30000 HY15000 HY03000 HYR20030 HYR10015 HY01500
103258-460EA 256.83 USD InStock
103258-460 103258-472 103258-470 103258-464 103258-462 103258-468 103258-466
HYPE-5™ Transfection Reagent, OZ Biosciences
Transfection Reagents
HYPE-5 Transfection Kit is dedicated to achieve High Yield Protein Expression in mammalian cells.
  • High yield recombinant protein production
  • Suitable for both HEK293 and CHO cells growing in suspension
  • Compatible with any synthetic or regular media used for protein production
  • Completely animal origin free
This Kit has been designed for maximum recombinant protein expression in HEK293 and CHO cells growing in suspension.
Delivery information: Shipped at room temperature
Packaging: Store Hype-5 Reagent at -20 °C and Hype-5 Blast at +4 °C
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty