Would you like to log in again?
STANZOIL® Chem-Ply™ Chemical Resistance Gloves, Mapa Professional
Supplier: Mapa Professionnel
STANZOIL®
413701 413709 413700
32917-264CS 444.39 USD InStock
32917-264 32917-266 32917-268
STANZOIL® Chem-Ply™ Chemical Resistance Gloves, Mapa Professional
Gloves
Ultra-high performance chemical protection gloves.
  • Broad chemical resistance (acids, aliphatic solvents, solvent mixtures), increased by the thickness of the material
  • Good grip, due to embossed texture
  • Easy to pull on and off, with a chlorinated interior
Two superimposed neoprene layers increase the mechanical and chemical resistance. The glove's red inner layer ensures that any wear is clearly visible, providing optimal safety for the user.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty