Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium hydroxide 1.0 M KOH (1.0 N) volumetric solution, Fluka™

Supplier: Honeywell Research Chemicals
Danger
Synonyms: Potassium hydrateCaustic potash

319376-500ML 319376-2L 319376-2L
BJ319376-500MLEA 71.27 USD InStock
BJ319376-500ML CABJ319376-2L BJ319376-2L
Potassium hydroxide 1.0 M KOH (1.0 N) volumetric solution, Fluka™
Potassium hydroxide


Formula: KOH
MW: 56.11 g/mol
Density: 1.05 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003553
CAS Number: 1310-58-3
UN: 1814
ADR: 8,II
Order Now
Specification Test Results
Appearance complying
Coloration complying
Clearness complying
Assay titr. 0.950 - 1.050 mol/l
Traceable to NIST SRM complying
BAM/EMPA reference material complying
BAM/EMPA Certificate Number complying
Carbonate (CO₃) Max. 0.01 mol/l

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty