Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Acetone ≥99.8% (by GC), CHROMASOLV™ for analysis of dioxins, furans and PCB, Riedel-de Haën™

Supplier: Honeywell Research Chemicals
Danger
Synonyms: 2-PropanoneDMKPropanoneDimethyl ketone

31062-2.5L
BJ31062-2.5LEA 99.25 USD InStock
BJ31062-2.5L
Acetone ≥99.8% (by GC), CHROMASOLV™ for analysis of dioxins, furans and PCB, Riedel-de Haën™
Acetone


Formula: CH₃COCH₃
MW: 58.08 g/mol
Boiling Pt: 55-57 °C (1.013 hPa)
Melting Pt: -95 °C
Density: 0.79 g/cm³ (20 °C)
Flash Pt: 14.3 %(V)
Storage Temperature: Ambient
MDL Number: MFCD00008765
CAS Number: 67-64-1
EINECS: 200-662-2
UN: 1090
ADR: 3,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/06/07/720607.jpg
Order Now
Specification Test Results
Refractive Index n20/D 1.359 (lit.)
Vapor Density 2 (vs air)
Assay (GC) Min. 99.8 %
Non-volatile matter Max. 0.0005 %
Water (Karl Fischer) Max. 0.2 %
Suitability for GC-MS of dioxines, furanes, PCPs complying

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty