Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Toluene ≥99.7%, CHROMASOLV™ for analysis of dioxins, furans and PCB, Riedel-de Haën™

Supplier: Honeywell Research Chemicals
Danger
Synonyms: MethacideBenzene, methylMethylbenzolToluolPhenylmethaneMethylbenzeneMethane, phenyl-

34413-2.5L
BJ34413-2.5LEA 156.73 USD InStock
BJ34413-2.5L
Toluene ≥99.7%, CHROMASOLV™ for analysis of dioxins, furans and PCB, Riedel-de Haën™
Toluene


Formula: C₆H₅CH₃
MW: 92.14 g/mol
Boiling Pt: 110.6 °C
Melting Pt: -95 °C
Density: 0.867 g/cm³ (20 °C)
Flash Pt: 7.1 %(V)
Storage Temperature: Ambient
MDL Number: MFCD00008512
CAS Number: 108-88-3
EINECS: 203-625-9
UN: 1294
ADR: 3,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/61/73/766173.jpg
Order Now
Specification Test Results
Assay (GC) Min. 99.7 %
Non-volatile matter Max. 0.0005 %
Water (Karl Fischer) Max. 0.02 %
Suit.f.GC-MS of dioxines,furanes,PCBs complying
Vapor Pressure 22 mmHg ( 20 °C)
Explosion Limit 7 %
Vapor Density 3.2 (vs air)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty