Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

n-Pentane ≥99% (by GC), CHROMASOLV™ AMD, Riedel-de Haën™

Supplier: Honeywell Research Chemicals
Danger
Synonyms: Pentane

34894-4X2.5L
BJ34894-2.5LCS 1129.28 USD InStock
BJ34894-2.5L
n-Pentane ≥99% (by GC), CHROMASOLV™ AMD, Riedel-de Haën™
n-Pentane
  • Solvents designed for high-performance thin-layer chromatography


Formula: H₃C(CH₂)₃CH₃
MW: 72.15 g/mol
Boiling Pt: 36-37 °C
Melting Pt: -131 °C
Density: 0.631 g/cm³ (15 °C)
Flash Pt: <-35 °C
Storage Temperature: Ambient
MDL Number: MFCD00009498
CAS Number: 109-66-0
EINECS: 203-692-4
UN: 1265
ADR: 3,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/81/67/738167.jpg
Order Now
Specification Test Results
Refractive Index n20/D 1.358 (lit.)
Assay (GC) Min. 99 %
Non-volatile matter Max. 0.0005 %
Water (Karl Fischer) Max. 0.005 %
Free acid (as CH₃COOH) Max. 0.0005 %
Transmittance at 200 nm Min. 20 %
Transmittance at 210 nm Min. 60 %
Transmittance at 220 nm Min. 85 %
Transmittance at 230 nm Min. 95 %
Transmittance from 250 nm Min. 99 %
Fluorescence (chinin) at 254 nm Max. 1 ppb
Fluorescence (chinin) at 365 nm Max. 1 ppb
for AMD technic complying
Vapor Density 2.48 (vs air)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty