Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Benzyl alcohol ≥99.9% (by GC), CHROMASOLV™ for headspace gas chromatography, Riedel-de Haën™

Supplier: Honeywell Research Chemicals
Warning
Synonyms: PhenylmethanolPhenylcarbinol

80708-1L
BJ80708-1LEA 447.56 USD InStock
BJ80708-1L
Benzyl alcohol ≥99.9% (by GC), CHROMASOLV™ for headspace gas chromatography, Riedel-de Haën™
Benzyl alcohol
Honeywell Chromasolv™ GC-headspace solvents:

  • Developed particularly for the identification and quantification of residual solvents in pharmaceuticals using GC-headspace methods described in monographs from the United States Pharmacopeia (USP) or the European Pharmacopoeia (Ph. Eur.) and following the International Conference on Harmonization of Technical Requirements for Registration of Pharmaceuticals for Human Use (ICH) guidelines
  • Very pure solvents with extremely low concentrations of the defined residual solvents


Formula: C₆H₅CH₂OH
MW: 108.14 g/mol
Boiling Pt: 205.3 °C (1.013 hPa)
Melting Pt: -15.4 °C
Density: 1.045 g/cm³ (20 °C)
Flash Pt: 100.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00004599
CAS Number: 100-51-6
EINECS: 202-859-9
https://us-legacy.vwr.com/stibo/web/std.lang.all/97/87/2299787.jpg
Order Now
Specification Test Results
Assay (GC) Min. 99.9 %
Refractive index (n 20/D) 1.538 - 1.541
Water (Karl Fischer) Max. 0.02 %
Acetophenone (GC) Max. 0.01 %
Benzaldehyde (GC) Max. 0.01 %
Suitability for the GC Headspace complying
Vapor Pressure 3.75 mmHg ( 77 °C)
Autoignition Temperature 817 °F
Vapor Density 3.7 (vs air)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty