Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hydrogen peroxide solution ≥35% (RT) stabilized, Purum p.a.

Supplier: Honeywell Research Chemicals
Danger

95299-2.5L
BJ95299-2.5LEA 232.97 USD InStock
BJ95299-2.5L
Hydrogen peroxide solution ≥35% (RT) stabilized, Purum p.a.
Hydrogen peroxide
  • Products with a guarantee certificate and/or suitable for the stated analytical application.


Formula: H₂O₂
MW: 34.01 g/mol
Melting Pt: -33 °C
Density: 1.110 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011333
CAS Number: 7722-84-1
UN: 2014
ADR: 5.1,II
Order Now
Specification Test Results
Assay 34.5 - 37.0 %
Density (D 20/4) 1.131 - 1.133
Free acid (as H₂SO₄) Max. 0.02 %
Calcium (Ca) Max. 10 ppm
Cadmium (Cd) Max. 5 ppm
Cobalt (Co) Max. 5 ppm
Chromium (Cr) Max. 5 ppm
Copper (Cu) Max. 5 ppm
Potassium (K) Max. 50 ppm
Iron (Fe) Max. 5 ppm
Magnesium (Mg) Max. 5 ppm
Manganese (Mn) Max. 5 ppm
Sodium (Na) Max. 200 ppm
Nickel (Ni) Max. 5 ppm
Lead (Pb) Max. 5 ppm
Zinc (Zn) Max. 5 ppm
Appearance (color) complying
Appearance (form) complying

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty