Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium hypochlorite 4.00 - 4.99% available chlorine in aqueous solution, Reagent Grade

Supplier: Honeywell Research Chemicals
Danger

239305-500ML 239305-5L
BJ239305-500MLEA 83.87 USD InStock
BJ239305-500ML BJ239305-5L
Sodium hypochlorite 4.00 - 4.99% available chlorine in aqueous solution, Reagent Grade
Sodium hypochlorite


Formula: ClNaO
MW: 74.44 g/mol
Melting Pt: –30…–20 °C
Density: 1.1 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011120
CAS Number: 7681-52-9
EINECS: 231-668-3
ADR: 8,III
Order Now
Specification Test Results
Assay of active chlorine 4 - 5 %
Vapor Pressure 17.5 mmHg ( 20 °C)
Composition available chlorine, 4.00-4.99%

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty