Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Acetone ≥99.9%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™

Supplier: Honeywell Research Chemicals
Danger
Synonyms: Dimethyl ketoneDimethylformaldehydeDMKPropanone2-Propanone

650501-4X4L 650501-1L
BJ650501-4LCS 1021.99 USD InStock
BJ650501-4L BJ650501-1L
Acetone ≥99.9%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Acetone


Formula: CH₃COCH₃
MW: 58.08 g/mol
Boiling Pt: 55-57 °C (1.013 hPa)
Melting Pt: -95 °C
Density: 0.79 g/cm³ (20 °C) (lit.)
Flash Pt: 14.3 %(V)
Storage Temperature: Ambient
MDL Number: MFCD00008765
CAS Number: 67-64-1
EINECS: 200-662-2
UN: 1090
ADR: 3,II
Order Now
Specification Test Results
Refractive Index n20/D 1.359 (lit.)
Purity (GC) Min. 99.90 %
GC (as heptachlor epoxide) Max. 10 ng/l
Water (Karl Fischer) Max. 0.5 %
Non-volatile matter Max. 0.0003 %
IR spectrum complying
Absorbance at 400 nm Max. 0.005
Absorbance at 375 nm Max. 0.005
Absorbance at 350 nm Max. 0.01
Absorbance at 340 nm Max. 0.06
Absorbance at 330 nm Max. 1.0
Fluorescence (chinin) at 365 nm Max. 1.0 ppb
Explosion Limit 13.2 %
Vapor Pressure 184 mmHg ( 20 °C)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty