Would you like to log in again?
Reservoir Bottle, WHEATON®, DWK Life Sciences
Supplier: DWK Life Sciences
264711
89012-098CS 699.22 USD InStock
89012-098
Reservoir Bottle, WHEATON®, DWK Life Sciences
Bottles Glass Bottles
Ideal as mobile phase or filtration reservoir.
  • UV protectant coating
  • Manufactured from WHEATON 33 low extractable borosilicate glass
  • Autoclavable
Safety coated glass bottles provide an added measure of protection.
Certifications: Conforms to USP Type I and ASTM E 438 Type I, Class A requirements.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty