Would you like to log in again?
Whatman™ Multicompressor Tray for Mini UniPrep G2 Syringeless HPLC Filter
Supplier: Whatman products (Cytiva)
Whatman™
MUPG2MCWT8 MUPG2MCWT8
89499-528EA 32.71 USD InStock
89499-528 CA89499-528
Whatman™ Multicompressor Tray for Mini UniPrep G2 Syringeless HPLC Filter
Presses
A Mini UniPrep G2 multicompressor tray allows you to preload eight samples, further increasing the time savings provided by Cytiva's Mini-UniPrep G2 syringeless filtration line.
  • A multicompressor streamlines syringeless filtration - preload in advance with an extra tray
  • Designed for Cytiva's Mini-UniPrep G2, which combines a borosilicate glass vial, plunger and attached filter membrane, and septum with cap
The borosilicate glass design of the Mini-UniPrep G2 minimizes the risk of leachables contaminating a sample, making it an excellent choice for pharmaceutical quality control (pharma QC) and water monitoring applications.
Browse the Whatman line from Cytiva's business to find the right syringeless HPLC filter and compressor for your application.
Whatman Mini UniPrep G2 can be used with a multicompressor that can process up to eight samples at one time. An extra tray allows you to prepare samples in advance, resulting in reduced overall filtration time.
The glass design prevents interference from hydrocarbon extractables that could be found in plastic.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty