Would you like to log in again?
SP Wilmad-LabGlass Short Path Distilling Apparatus with Receiving Flasks, SP Industries
Supplier: SP Industries
LG-6315-098
14205-382EA 1807.34 USD InStock
14205-382
SP Wilmad-LabGlass Short Path Distilling Apparatus with Receiving Flasks, SP Industries
Distillation Components Distillation Sets
This borosilicate glass short path distilling head apparatus has a compact design to minimize product loss.
  • The thermometer bulb and stem serve as the column packing
A Standard Taper 10/18 outer joint accepts thermometers calibrated for 75 mm immersion. The distributing adapter consists of a single Standard Taper 24/40 outer joint and three Standard Taper 24/40 inner joints to accept three 50 mL round bottom receivers. The receivers are held in place with plastic clamps.
Ordering information: Complete unit comes with the following components; LG-6315-100 Distilling Head; LG-6315-102 Distributing Adapter 100 mL; LG-7291-234 2-Neck Flask 250 mL Standard Taper 24/40; LK-181-100 Bleed-in Adapter; LG-7220-140 Receiving Flask 50 mL (3 req’d); ML-1820-730 Thermometer –10 to +250 °C in 1 °C 75 mm immersion; LG-1004-108 Plastic Clamps Size 24; and LG-1453-106 Thermometer Adapter Standard Taper 24/40.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty