Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
SP Wilmad-LabGlass Short Path Distilling Apparatus with Separable Adapter Tubes, SP Industries
Supplier: SP Industries
ML-905-700 ML-905-704 ML-905-702
14209-740EA 329.24 USD InStock
14209-740 14205-842 14209-082
SP Wilmad-LabGlass Short Path Distilling Apparatus with Separable Adapter Tubes, SP Industries
Distillation Components Distillation Sets
This borosilicate glass short path distilling head apparatus has the distilling portion, condenser, and vacuum connection sealed close together to reduce travel path of distillate.
  • A standard taper 10/18 outer joint accepts the thermometer
The distributing adapter consists of a single Standard Taper 12/18 outer joint and three Standard Taper 12/18 inner joints ready to accept 3, 3 mL receivers.
Ordering information: Complete unit comes with the following components; ML-905-702 Distilling Head; ML-905-704 Distributing Adapter Standard Taper 12/18; L-905-706 Receiver 3 mL in 0.2 mL Standard Taper 12/18 (3 req’d); ML-905-800 Thermometer –10 to +250 °C in 1 °C Standard Taper 10/18; ML-178-700 Gas Inlet Adapter; and ML-1170-734 Distilling Flask 10 mL Standard Taper 14/20.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty