Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
Myokinase (from Yeast)
Supplier: EMD Millipore
475941-1KU
80603-218EA 140.7 USD InStock
80603-218
Myokinase (from Yeast)
Enzymes
Myokinase is native of yeast and reversibly transfers phosphate from ATP to AMP, thus forming two molecules of ADP
  • Lyophilized from potassium phosphate buffer
  • Hygroscopic in nature
Synonyms: Adenylate Kinase; ATP:AMP Phosphotransferase
The enzyme is specific for adenine nucleotides and catalyzes the reaction only in the presence of a divalent metal ion (e.g. Mg2+, Ca2+, Co2+, Mn2+, or Ni2+).
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty